Anti-Cytokeratin 5 Mouse Monoclonal Antibody [clone: XM26]
Supplier: Diagnostic Biosystems
MOB361-01-RUO MOB361-05-RUO MOB361-RUO
CA76633-758EA 144.79 CAD
CA76633-758 CA76633-760 CA76633-762
Anti-Cytokeratin 5 Mouse Monoclonal Antibody [clone: XM26]
Antibodies
This antibody reacts with cytokeratin 5 intermediate filament protein. Basal cells of human epidermis express acidic cytokeratin 14 and basic cytokeratin 5. Cytokeratin 5 is a 58 kDa protein, which is closely related to cytokeratin 6. Cytokeratin 5 and 6 are expressed in basal cell epithelioma, basal cells of prostate, urothelium, vagina, squamous cell carcinomas of skin, tongue, epiglottis and rectal-anal region. Cytokeratin 5 can be useful in the distinction of mesotheliomas from most adenocarcinomas.