Anti-Rad51 Mouse Monoclonal Antibody [clone: 51RAD01]
Supplier: Diagnostic Biosystems
MOB324-01-RUO MOB324-05-RUO MOB324-RUO
CA76633-680EA 185.68 CAD
CA76633-680 CA76633-682 CA76633-684
Anti-Rad51 Mouse Monoclonal Antibody [clone: 51RAD01]
Antibodies
This antibody is specific to a 37 to 43 kDa protein known as Rad51. Rad51 is similar to RecA. Rad51 promotes homologous pairing and strand exchange within a regular nucleoprotein filament. Rad51 has been suggested to be involved in homologous recombination and in cell proliferation regulation.