Anti-Myogenin Mouse Monoclonal Antibody [clone: F5D]
Supplier: Diagnostic Biosystems
MOB322-01-RUO MOB322-05-RUO MOB322-RUO
CA76633-668EA 161.82 CAD
CA76633-668 CA76633-670 CA76633-672
Anti-Myogenin Mouse Monoclonal Antibody [clone: F5D]
Antibodies
This antibody is specific to 34 kDa protein known as myogenin. Myogenin is a member of family of myogenic regulatory genes which includes MyoD, myf5 and MRF4. These genes encode a set of transcription factors which are essential for muscle development. Expression of myogenin is restricted to cells of skeletal muscle origin.