Anti-EBV Cocktail Mouse Monoclonal Antibody [clone: CS1, CS2, CS3, CS4]
Supplier: Diagnostic Biosystems
MOB194-01-RUO MOB194-05-RUO MOB194-RUO
CA76633-418EA 144.79 CAD
CA76633-418 CA76633-420 CA76633-422
Anti-EBV Cocktail Mouse Monoclonal Antibody [clone: CS1, CS2, CS3, CS4]
Antibodies
This antibody is specific to 60 kDa latent membrane protein (LMP-1) encoded by the BNLF1 gene of the EBV. Each clone reacts with different epitopes on the hydrophilic C-terminus of the cytoplasmic domain of LMP-1. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis.