Anti-Cytokeratin AE1/AE3 Mouse Monoclonal Antibody [clone: AE1+AE3]
Supplier: Diagnostic Biosystems
MOB190-RUO MOB190-01-RUO MOB190-05-RUO
CA76633-416EA 645.56 CAD
CA76633-416 CA76633-412 CA76633-414
Anti-Cytokeratin AE1/AE3 Mouse Monoclonal Antibody [clone: AE1+AE3]
Antibodies
This AE1/AE3 antibody cocktail recognizes most of the acidic (Type I) keratins as well as basic (Type II). It recognizes keratin polypeptides of 40, 48, 50, 52, 54, 56.5, 58, 59 and 64 to 67 kDa. In keratinized epidermis, 50 kDa keratin is present in the basal layer, while 56.5 kDa keratin is present in suprabasal layers. 58 kDa keratin is present in the basal and suprabasal layers, while 65 to 67 kDa keratin is present in the cells above the basal layers. This antibody can be used to stain distinct cell layers in an epithelial tissue and various neoplasms of epithelial origin.