Anti-Cytokeratin 14 Mouse Monoclonal Antibody [clone: LL002]
Supplier: Diagnostic Biosystems
MOB186-05-RUO MOB186-RUO MOB186-01-RUO
CA76633-396EA 463.29 CAD
CA76633-396 CA76633-398 CA76633-394
Anti-Cytokeratin 14 Mouse Monoclonal Antibody [clone: LL002]
Antibodies
This antibody is specific to 50 kDa cytokeratin protein designated cytokeratin 14. Cytokeratin 14 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 15. This antibody can be used to distinguish stratified epithelial cells from simple epithelial cells.