Anti-Cytokeratin 20 Mouse Monoclonal Antibody [clone: Ks20.8]

Supplier: Diagnostic Biosystems
MOB123-05-RUO MOB123-RUO MOB123-01-RUO
CA76633-336EA 637.02 CAD
CA76633-336 CA76633-338 CA76633-334
Anti-Cytokeratin 20 Mouse Monoclonal Antibody [clone: Ks20.8]
Antibodies
Cytokeratin 20 is less acidic than the other type I cytokeratins and is of particular interest because of its restricted range of expression. This antibody reacts with cytokeratin polypeptide of 46 kDa in immunoblot analyses. Studies have shown that a marked difference exists in the expression of Cytokeratin 20 among various carcinomas.
Order Now

SPECIFICATIONS

Antigen name Cytokeratin 20
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Ks20.8
Reactivity Human
Host Mouse
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR