Anti-CD79a B-cell Mouse Monoclonal Antibody [clone: HM57]
Supplier: Diagnostic Biosystems
MOB118-01-RUO MOB118-05-RUO MOB118-RUO
CA76633-322EA 228.26 CAD
CA76633-322 CA76633-324 CA76633-326
Anti-CD79a B-cell Mouse Monoclonal Antibody [clone: HM57]
Antibodies
This antibody reacts specifically with B cells at the pre B cell stage. It stains a majority of acute leukemias of precursor B cell type, B cell lymphomas and some myelomas. The mb-1 polypeptide along with B29 polypeptide forms an antigen receptor complex on the B cell, performing signal transduction and phosphorylation functions.