Anti-Lambda light chain Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP027-01-RUO RP027-05-RUO RP027-RUO
CA76635-000EA 127.93 CAD
CA76635-000 CA76635-002 CA76635-004
Anti-Lambda light chain Rabbit Polyclonal Antibody
Antibodies
This antibody does not show any cross-reactivity with kappa light chain. It stains B cell follicles in human lymphoid tissues. The mantle zone shows a mosaic pattern of labeling while the germinal centers show a coarse meshwork pattern of staining.