Anti-Kappa light chain Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP025-01-RUO RP025-05-RUO RP025-RUO
CA76634-988EA 127.93 CAD
CA76634-988 CA76634-990 CA76634-992
Anti-Kappa light chain Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with free as well as bound kappa light chains. Contaminating antibodies have been removed by solid phase adsorption.