Anti-Cytokeratin wide spectrum Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
RP010-01-RUO RP010-05-RUO RP010-RUO
CA76634-922EA 186.63 CAD
CA76634-922 CA76634-924 CA76634-926
Anti-Cytokeratin wide spectrum Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with cytokeratins of 58, 56, 52, 60, 51, 48 and 68 kDa MW. This antibody is well suited for the staining of a broad spectrum of human keratins.
Order Now

SPECIFICATIONS

Antigen name Cytokeratin wide spectrum Cytokeratin wide spectrum
Environmentally Preferable
Antibody type Primary Primary
Clonality Polyclonal Polyclonal
Conjugation Unconjugated Unconjugated
Reactivity Human Human
Host Rabbit Mouse
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR