Anti-Chromogranin A Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP008-01-RUO RP008-05-RUO RP008-RUO
CA76634-916EA 173.07 CAD
CA76634-916 CA76634-918 CA76634-920
Anti-Chromogranin A Rabbit Polyclonal Antibody
Antibodies
Chromogranin A is a 439 amino acid protein present in secretory granules of a wide variety of endocrine cells and neurons. A positive staining is seen in the secretory granules of parathyroid, adrenal medulla, anterior pituitary gland and Langerhans islets of pancreas. This antibody stains a variety of neuroendocrine tumors.