Anti-HER2/neu (c-erb-2) Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP006-01-RUO RP006-05-RUO RP006-RUO
CA76634-904EA 127.93 CAD
CA76634-904 CA76634-906 CA76634-908
Anti-HER2/neu (c-erb-2) Rabbit Polyclonal Antibody
Antibodies
This antibody recognizes a c-erbB-2 protein, which is a receptor tyrosine kinase of the c-erbB family. It is closely related in structure to the epidermal growth factor receptor. c-erbB-2 oncoprotein is detectable in a subset of breast and other adenocarcinomas, as well as transitional cell carcinomas. In the case of breast cancer, expression determined by immunohistochemistry has been shown to be associated with poor prognosis.