Anti-CD5 Mouse Monoclonal Antibody [clone: 54/F6]
Supplier: Diagnostic Biosystems
MOB589-01-RUO MOB589-05-RUO MOB589-RUO
CA76634-216EA 183.98 CAD
CA76634-216 CA76634-218 CA76634-220
Anti-CD5 Mouse Monoclonal Antibody [clone: 54/F6]
Antibodies
This antibody recognizes CD5, which is a transmembrane protein, is found on 95% of thymocytes and 72% of peripheral blood lymphocytes. In lymph nodes, the main reactivity is observed in T cell areas. CD5 is expressed by many T cell leukemia, lymphomas, and activated T cells. Occasionally, CD5 antigen is also expressed on a subset of B cells. Mantle cell lymphomas (same as diffuse centrocytic lymphomas) are CD5(+) while the follicle center cell lymphoma are CD5(-).