Anti-SOX10 Mouse Monoclonal Antibody [clone: 20B7]
Supplier: Diagnostic Biosystems
MOB565-01-RUO MOB565-05-RUO MOB565-RUO
CA76634-138EA 144.79 CAD
CA76634-138 CA76634-140 CA76634-142
Anti-SOX10 Mouse Monoclonal Antibody [clone: 20B7]
Antibodies
SOX10 is widely expressed in normal human tissues including melanocytes and breast tissue. SOX10 is also an important marker in malignant tumors such as melanoma, breast carcinoma, gliomas, and benign tumors such as schwannomas.SOX-10 is considered as a very reliable marker for recognizing residual desmoplastic melanomas. In normal tissues, it is expressed in Schwann cells, melanocytes and myoepithelial cells of salivary, bronchial and mammary glands. SOX-10 expression is also observed in mast cells.