Anti-Melanoma pan cocktail Mouse Monoclonal Antibody [clone: HMB45 + A103 + T311]
Supplier: Diagnostic Biosystems
MOB428-01-RUO MOB428-05-RUO MOB428-RUO
CA76633-880EA 253.8 CAD
CA76633-880 CA76633-882 CA76633-884
Anti-Melanoma pan cocktail Mouse Monoclonal Antibody [clone: HMB45 + A103 + T311]
Antibodies
The HMB45 clone reacts with a neuraminidase-sensitive oligosaccharide side chain of a glycoconjugate present in immature melanosomes. The HMB45-reactive antigen is present in cutaneous melanocytes, prenatal and infantile retinal pigment epithelium and melanoma cells and is thought to be oncofetal in nature. This antibody has been shown to label the majority of melanomas. Clone A103 recognizes a protein of 20 kDa, identified as MART-1 (melanoma antigen recognized by T cells-1) or Melan-A. Melan-A is a useful addition to melanoma panels as it is apparently specific for melanocytic lesions. Studies have also shown that MART-1 is more sensitive than HMB45 when labeling metastatic melanomas. Tyrosinase is a key enzyme involved in the initial stages of melanin biosynthesis. Studies have shown tyrosinase to be a more sensitive marker when compared to HMB45 and MART-1. It has been shown to label a higher percentage of desmoplastic melanomas than HMB45. The combination of HMB45, MART-1 and tyrosinase make this triple antibody cocktail a first-order pan melanoma screener.