Anti-Hepatocyte Mouse Monoclonal Antibody [clone: OCH1E5]
Supplier: Diagnostic Biosystems
MOB426-01-RUO MOB426-05-RUO MOB426-RUO
CA76633-874EA 144.79 CAD
CA76633-874 CA76633-876 CA76633-878
Anti-Hepatocyte Mouse Monoclonal Antibody [clone: OCH1E5]
Antibodies
Hepatoblastoma is the most common primary tumor of the liver in children. The use of specific hepatocyte markers and also of α fetoprotein or carcinoembryonic antigen are useful for the identification of normal and malignant fetal hepatocytes. This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.