Anti-Factor VIII Mouse Monoclonal Antibody [clone: F8/86]
Supplier: Diagnostic Biosystems
MOB196-01-RUO MOB196-05-RUO MOB196-RUO
CA76633-424EA 144.79 CAD
CA76633-424 CA76633-426 CA76633-428
Anti-Factor VIII Mouse Monoclonal Antibody [clone: F8/86]
Antibodies
This antibody reacts with Von Willebrand factor in endothelial cells. It also reacts with megakaryocytes in human bone marrow.