Anti-Smad4 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP173-01-RUO RP173-05-RUO RP173-RUO
CA76635-186EA 99.33 CAD
CA76635-186 CA76635-188 CA76635-190
Anti-Smad4 Rabbit Polyclonal Antibody
Antibodies
SMADs are members of the MAD-related family of molecules. MAD-related proteins are a family of intracellular proteins that are essential components in the signaling pathways of the serine/threonine kinase receptors of the transforming growth factor beta superfamily. Alterations in the SMAD4 gene was primarily discovered in pancreatic cancer (duct adenocarcinoma) but occur in a variety of cancers such as colorectal cancer, gastric cancer, prostate cancer, melanomas, head and neck cancers and many others, though with higher frequencies in gastrointestinal tract cancers. Loss of SMAD4More Product Information