Anti-Uroplakin III (AU1) Mouse Monoclonal Antibody [clone: AU1]
Supplier: Diagnostic BioSystems
PDM594-RUO PDM594-RUO
CA76634-594EA 506.8 CAD
CA76634-594 76634-594
Anti-Uroplakin III (AU1) Mouse Monoclonal Antibody [clone: AU1]
Antibodies
Uroplakins (UPs) are a family of transmembrane proteins (UPs Ia, Ib, II and III) that are specific differentiation products of urothelial cells. In nonneoplastic mammalian urothelium, UPs are expressed in the luminal surface plasmalemma of superficial (umbrella) cells, forming complexes of 16nm crystalline particles. UPIII is specific for tumors of urothelial origin and, when used in combination with other markers, can aid in the diagnosis of primary and metastatic tumors.