Anti-BRCA1 Mouse Monoclonal Antibody [clone: GLK-2]
Supplier: Diagnostic Biosystems
MOB424-01-RUO MOB424-05-RUO MOB424-RUO
CA76633-868EA 148.19 CAD
CA76633-868 CA76633-870 CA76633-872
Anti-BRCA1 Mouse Monoclonal Antibody [clone: GLK-2]
Antibodies
BRCA1 recognizes a 22kDa protein. This antibody stains 94% of ovarian carcinomas with a mutation in exon 11. However, staining was absent in 100% of tumors with mutations other than exon 11.