Anti-S-100 Mouse Monoclonal Antibody [clone: 4C4.9]

Supplier: Diagnostic Biosystems
MOB377-01-RUO MOB377-05-RUO MOB377-RUO
CA76633-778EA 144.79 CAD
CA76633-778 CA76633-780 CA76633-782
Anti-S-100 Mouse Monoclonal Antibody [clone: 4C4.9]
Antibodies
This antibody is specific against an epitope located on the β chain (i.e., in S-100a and S-100b) but not on the α- chain of S-100 (i.e., in S-100a and S-100ao). This antibody can be used to localize S-100a and S-100b in various tissue sections.
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name S-100
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 4C4.9
Reactivity Human
Host Mouse
Isotype IgG2a

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR