Anti-CD1A Cortical Thymocytes Mouse Monoclonal Antibody [clone: O10]
Supplier: Diagnostic Biosystems
MOB363-05-RUO MOB363-RUO
CA76633-768EA 720.5 CAD
CA76633-768 CA76633-770
Anti-CD1A Cortical Thymocytes Mouse Monoclonal Antibody [clone: O10]
Antibodies
This antibody reacts with a 49 kDa protein. At least five CD1 genes are identified (CD1a, b, c, d, and e). This antibody reacts with cortical thymocytes, Langerhans cells in epidermis, dendritic cells of dermis and Langerhans cells of mucosa of tonsil. This antibody may be useful in the characterization of leukemias and lymphomas.