Anti-Laminin Mouse Monoclonal Antibody [clone: 4C7]
Supplier: Diagnostic Biosystems
MOB202-01-RUO MOB202-05-RUO MOB202-RUO
CA76633-436EA 161.82 CAD
CA76633-436 CA76633-438 CA76633-440
Anti-Laminin Mouse Monoclonal Antibody [clone: 4C7]
Antibodies
This antibody reacts with the terminal globular domain of the A chain. The staining with this antibody is sensitive to fixation of paraffin tissues and use of proteolytic enzymatic treatment.