Anti-Hairy Cell Leukemia Mouse Monoclonal Antibody [clone: DBA.44]
Supplier: Diagnostic Biosystems
MOB200-01-RUO MOB200-05-RUO MOB200-RUO
CA76633-430EA 155.01 CAD
CA76633-430 CA76633-432 CA76633-434
Anti-Hairy Cell Leukemia Mouse Monoclonal Antibody [clone: DBA.44]
Antibodies
This antibody reacts with a subset of B-lymphocytes localized in the follicular mantle zone. It reacts with 97% of hairy cell leukemia cases. This antibody shows strong positive staining of about 35% of cases of high grade B cell lymphomas.