Anti-Herpes Simplex Virus II (HSV II) Mouse Monoclonal Antibody [clone: DBM15.69]
Supplier: Diagnostic Biosystems
MOB542-01-RUO MOB542-05-RUO MOB542-RUO
CA76634-030EA 165.22 CAD
CA76634-030 CA76634-032 CA76634-034
Anti-Herpes Simplex Virus II (HSV II) Mouse Monoclonal Antibody [clone: DBM15.69]
Antibodies
This antibody reacts with herpes simplex virus type 2. It is specific for the viral glycoprotein D (gD) protein.