Anti-Adrenocorticotropic Hormone AH26 Mouse Monoclonal Antibody [clone: AH26]
Supplier: Diagnostic Biosystems
MOB244-01-RUO MOB244-05-RUO MOB244-RUO
CA76633-510EA 144.79 CAD
CA76633-510 CA76633-512 CA76633-514
Anti-Adrenocorticotropic Hormone AH26 Mouse Monoclonal Antibody [clone: AH26]
Antibodies
This antibody is specific to an epitope on the N-terminus of ACTH. This antibody stains corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas.