Anti-PSAP Mouse Monoclonal Antibody [clone: PASE/4LJ]
Supplier: Diagnostic Biosystems
MOB085-01-RUO MOB085-05-RUO MOB085-RUO
CA76633-256EA 144.79 CAD
CA76633-256 CA76633-258 CA76633-260
Anti-PSAP Mouse Monoclonal Antibody [clone: PASE/4LJ]
Antibodies
This antibody is specific to human prostatic acid phosphatase. It reacts with the glandular epithelium of normal and hyperplastic prostate and with prostate adenocarcinoma.