Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]

Supplier: Diagnostic Biosystems
MOB083-01-RUO MOB083-05-RUO MOB083-RUO
CA76633-250EA 156.7 CAD
CA76633-250 CA76633-252 CA76633-254
Anti-PCNA Mouse Monoclonal Antibody [clone: PC10]
Antibodies
PCNA is essential for cellular DNA synthesis. This antibody reacts with proliferating cells in a wide range of normal tissues. The significance of PCNA immunoreactivity in tumors in not fully established. However, in non-Hodgkin lymphoma a linear relation between Ki-67 and PCNA staining has been demostrated.
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name PCNA
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone PC10
Reactivity Human
Host Mouse
Isotype IgG2a kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR