Anti-Heat Shock Protein 70 Mouse Monoclonal Antibody [clone: W27]
Supplier: Diagnostic Biosystems
MOB269-01-RUO MOB269-05-RUO MOB269-RUO
CA76633-552EA 165.22 CAD
CA76633-552 CA76633-554 CA76633-556
Anti-Heat Shock Protein 70 Mouse Monoclonal Antibody [clone: W27]
Antibodies
This antibody is specific to constitutive (HSP73) and inducible (HSP72) form of HSP70. HSP70 is found to be associated with steroid receptors, actin, P53, polyoma T antigen, nucleotides and other known proteins. HSP70 has been shown to be involved in protective roles against thermal stress, cytotoxic drugs and other damaging conditions.