Anti-Cadherin Pan Mouse Monoclonal Antibody [clone: CH-19]
Supplier: Diagnostic Biosystems
MOB228-01-RUO MOB228-05-RUO MOB228-RUO
CA76633-468EA 144.79 CAD
CA76633-468 CA76633-470 CA76633-472
Anti-Cadherin Pan Mouse Monoclonal Antibody [clone: CH-19]
Antibodies
This antibody reacts with pan cadherin with a distinct 135 kDa band from a wide variety of tissues. Cadherins are members of a multigene family of single chain glycoprotein receptors mediating Ca ²⁺ dependent cell-cell adhesion.