Anti-Glycophorin C Mouse Monoclonal Antibody [clone: Ret40f]

Supplier: Diagnostic Biosystems
MOB067-01-RUO MOB067-05-RUO MOB067-RUO
CA76633-212EA 211.23 CAD
CA76633-212 CA76633-214 CA76633-216
Anti-Glycophorin C Mouse Monoclonal Antibody [clone: Ret40f]
Antibodies
This antibody is specific to β sialoglycophorin also known as glycophorin C. Glycophorin C is found on the human erythrocyte membrane and is one of four sialic acid-rich polypeptides.
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Glycophorin C
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Ret40f
Reactivity Human
Host Mouse
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR