Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: PN-15]
Supplier: Diagnostic Biosystems
MOB465-01-RUO MOB465-05-RUO MOB465-RUO
CA76633-964EA 144.79 CAD
CA76633-964 CA76633-966 CA76633-968
Anti-Renal Cell Carcinoma Mouse Monoclonal Antibody [clone: PN-15]
Antibodies
This antibody recognizes a 200 kDa glycoprotein (GP200) localized in the brush border of the proximal renal tebules and the luminal surface of Bowman’s capsule. GP200 was expressed by 93% of primary and 84% of metastatic renal carcinomas. GP200 glycoprotein is also localized in the breast lobules and ducts, parathyroid glands and epididymis.