Anti-Cytokeratin 5 and 14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Supplier: Diagnostic Biosystems
MOB433-01-RUO MOB433-05-RUO MOB433-RUO
CA76633-898EA 144.79 CAD
CA76633-898 CA76633-900 CA76633-902
Anti-Cytokeratin 5 and 14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Antibodies
This antibody is specific to 50 kDa cytokeratin protein designated cytokeratin 14. Cytokeratin 14 belongs to the type A (acidic) subfamily of low molecular weight cytokeratins and exists in combination with cytokeratin 15. This antibody can be used to distinguish stratified epithelial cells from simple epithelial cells.