Anti-E2F-1 transcription factor Mouse Monoclonal Antibody [clone: KH95]
Supplier: Diagnostic BioSystems
MOB381-RUO MOB381-01-RUO MOB381-01 MOB381-05-RUO MOB381-05 MOB381
CA76633-794EA 1173.45 CAD
CA76633-794 76633-794 CA76633-792 76633-792 CA76633-790 76633-790
Anti-E2F-1 transcription factor Mouse Monoclonal Antibody [clone: KH95]
Antibodies
This antibody reacts with a protein of 60 kDa. E2Fs are DNA-binding proteins, which associate with negative regulators, such as the retinoblastoma P107 protein, resulting in an altered rate of gene transcription. E2F-1 is proposed to be involved in several cellular processes that range from tumor suppression, cell progression and oncogenesis. E2F-1 over expression can also drive cells into apoptosis.