Anti-Calponin Mouse Monoclonal Antibody [clone: CALP]
Supplier: Diagnostic Biosystems
MOB345-01-RUO MOB345-05-RUO MOB345-RUO
CA76633-716EA 190.77 CAD
CA76633-716 CA76633-718 CA76633-720
Anti-Calponin Mouse Monoclonal Antibody [clone: CALP]
Antibodies
This antibody reacts with a 34 kDa protein known as calponin. Calponin, a calmodulin, binds tropomyosin and F-actin and is thought to be involved in the regulation of smooth muscle contraction. Expression of calponin is restricted to smooth muscle cells. Two isoforms of calponin exist whose molecular weights are 34 kDa and 29 kDa. 29 kDa isoform is primarily restricted to the muscle of the urogenital tract.