Anti-Glial Fibrillary Acidic Protein (GFAP) Mouse Monoclonal Antibody [clone: GA5]
Supplier: Diagnostic Biosystems
MOB064-01-RUO MOB064-05-RUO MOB064-RUO
CA76633-200EA 144.79 CAD
CA76633-200 CA76633-202 CA76633-204
Anti-Glial Fibrillary Acidic Protein (GFAP) Mouse Monoclonal Antibody [clone: GA5]
Antibodies
This antibody reacts with the 52 kDa intermediate filament protein GFAP in brain and spinal cord. It labels some astrocytes and some CNS ependymal cells but not oligodendrocytes or neurons. This antibody does not react with other intermediate filament proteins.