Anti-CD20 B cell Mouse Monoclonal Antibody [clone: L26]
Supplier: Diagnostic Biosystems
MOB004-01-RUO MOB004-05-RUO MOB004-RUO
CA76633-092EA 144.79 CAD
CA76633-092 CA76633-094 CA76633-096
Anti-CD20 B cell Mouse Monoclonal Antibody [clone: L26]
Antibodies
This antibody is specific to a 33 kDa polypeptide present on the majority of B cells in peripheral blood and lymphoid tissue. No reactivity with other hematopoietic cells has been observed.