Anti-Caspase 8 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP097-05-RUO RP097-RUO RP097-01-RUO
CA76635-116EA 621.59 CAD
CA76635-116 CA76635-118 CA76635-114
Anti-Caspase 8 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 55 kDa protein. Caspase 8 has an N-terminal domain with sequence homology to the death effector domain (DED) of FADD. Caspase 8 binds to DED of FADD. Caspase 8 is activated by association with Fas/FADD death-inducing signaling complex to release two active subunits, P18 and P10 into the cytosol, where they activate other caspases amplifying the apoptotic signal.