Anti-Caspase 3 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP096-01-RUO RP096-05-RUO RP096-RUO
CA76635-108EA 153.51 CAD
CA76635-108 CA76635-110 CA76635-112
Anti-Caspase 3 Rabbit Polyclonal Antibody
Antibodies
This antibody is specific to a 32 kDa protein, which is identified as Caspase 3. Caspase 3 is associated with induction of apoptosis. Caspase 3 is ubiquitously expressed and is synthesized as an inactive proenzyme.