Anti-Beta-Catenin Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
RP080-01-RUO RP080-05-RUO RP080-RUO
CA76635-102EA 171.57 CAD
CA76635-102 CA76635-104 CA76635-106
Anti-Beta-Catenin Rabbit Polyclonal Antibody
Antibodies
The catenins are structurally related cytoplasmic proteins which have been classified as alpha (α), beta (β), and gamma (γ), these proteins can bind to the highly conserved, intracellular cytoplasmic tail of E-cadherin. Together, the catenin/cadherin complexes play an important role mediating cellular adhesion. β-catenin associates with the cytoplasmic portion of E-cadherin, which is necessary for the function of E-cadherin as an adhesion molecule.