Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: AA1]
Supplier: Diagnostic Biosystems
MOB347-01-RUO MOB347-05-RUO MOB347-RUO
CA76633-728EA 165.22 CAD
CA76633-728 CA76633-730 CA76633-732
Anti-Mast Cell Tryptase Mouse Monoclonal Antibody [clone: AA1]
Antibodies
This antibody reacts with mast cells distributed in skin, synovium, lung, and heart. This antibody does not bind with any other cell type. Mast cells contain a number of preformed chemicals mediators such as histamine, chymase, carboxypeptidase, and proteolytic tryptase. Human mast cell tryptase is considered to be an important marker of mast cell activation and is an important mediator of inflammation.