Anti-ERCC1 Mouse Monoclonal Antibody [clone: 8F1]
Supplier: Diagnostic Biosystems
MOB336-01-RUO MOB336-05-RUO MOB336-RUO
CA76633-698EA 185.68 CAD
CA76633-698 CA76633-700 CA76633-702
Anti-ERCC1 Mouse Monoclonal Antibody [clone: 8F1]
Antibodies
This antibody reacts with a 33-36 kDa protein known as ERCC1 (excision repair cross complementing) polypeptide. ERCC1 is required for nucleotide excision repair of damaged DNA and is homologous to RAD10. In mammalian cells, XPG cleaves 3’ of the DNA lesion while ERCC1-XPF complex makes the 5’ incision.