Anti-MLH-1 Mouse Monoclonal Antibody [clone: G168-15]
Supplier: Diagnostic Biosystems
MOB430-01-RUO MOB430-05-RUO MOB430-RUO
CA76633-892EA 163.51 CAD
CA76633-892 CA76633-894 CA76633-896
Anti-MLH-1 Mouse Monoclonal Antibody [clone: G168-15]
Antibodies
The G168-15 antibody recognizes human and mouse MLH1 (80 to 85 kDa). The repair of mismatch DNA is essential to maintaining the integrity of genetic information over time. An alteration of microsatellite repeats is the result of slippage owing to strand misalignment during DNA replication and is referred to as microsatellite instability (MSI). These defects in DNA repair pathways have been related to human carcinogenesis. The importance of mismatch repair genes became apparent with the identification of the genetic basis for hereditary nonpolyposis colon cancer (HNPC). MSH-2 is involved in the initial cognition of mismatch nucleotides during the replication mismatch repair process. It is thought that after MSH2 binds to a mismatched DNA duplex it is joined by a heterodimer of MLH1 and PMSH, which together help facilitate the later steps in mismatch repair.