Anti-Myosin Skeletal (Fast) Mouse Monoclonal Antibody [clone: MY32]
Supplier: Diagnostic Biosystems
MOB207-01-RUO MOB207-05-RUO MOB207-RUO
CA76633-442EA 144.79 CAD
CA76633-442 CA76633-444 CA76633-446
Anti-Myosin Skeletal (Fast) Mouse Monoclonal Antibody [clone: MY32]
Antibodies
This antibody recognizes heavy chain of human skeletal myosin. It does not stain cardiac or smooth muscle myosin. This antibody stains the fast twitch (type II) isomyosin molecule.