Anti-Epithelial Antigen Mouse Monoclonal Antibody [clone: VU-1D9]
Supplier: Diagnostic Biosystems
MOB062-05-RUO MOB062-RUO MOB062-01-RUO
CA76633-196EA 453.07 CAD
CA76633-196 CA76633-198 CA76633-194
Anti-Epithelial Antigen Mouse Monoclonal Antibody [clone: VU-1D9]
Antibodies
This antibody reacts with two glycoproteins of 34 and 49 kDa present on the epithelial cells. It stains with simple epithelia and basal layers of stratified non-keratinized squamous epithelium.