Anti-Cytokeratin HMW Mouse Monoclonal Antibody [clone: 34bE12]
Supplier: Diagnostic Biosystems
MOB059-01-RUO MOB059-05-RUO MOB059-RUO
CA76633-182EA 144.79 CAD
CA76633-182 CA76633-184 CA76633-186
Anti-Cytokeratin HMW Mouse Monoclonal Antibody [clone: 34bE12]
Antibodies
This antibody recognizes keratin polypeptides of 68, 58, 56.5 and 50 kDa in the stratum corneum. The antibody reacts with squamous, ductal and other complex epithelia. It stains adenocarcinomas, breast, pancreas, bile duct, salivary gland and transitional cell carcinomas.