Anti-Perforin Mouse Monoclonal Antibody [clone: 5B10]
Supplier: Diagnostic Biosystems
MOB555-01-RUO MOB555-05-RUO MOB555-RUO
CA76634-084EA 172.05 CAD
CA76634-084 CA76634-086 CA76634-088
Anti-Perforin Mouse Monoclonal Antibody [clone: 5B10]
Antibodies
This antibody is specific to a 70 kDa protein, which is a potent cytolytic pore-forming protein. Perforin is a specific marker of functionally active cytotoxic T-lymphocytes and natural killer cells. Perforin mediates cytolysis of target cells by membrane damage and apoptosis.