Anti-Myeloperoxidase Mouse Monoclonal Antibody [clone: 59A5]

Supplier: Diagnostic Biosystems
MOB551-01-RUO MOB551-05-RUO MOB551-RUO
CA76634-072EA 144.79 CAD
CA76634-072 CA76634-074 CA76634-076
Anti-Myeloperoxidase Mouse Monoclonal Antibody [clone: 59A5]
Antibodies
Myeloperoxidase metabolizes most of the hydrogen peroxide generated by activated phagocytes. This antibody stains strongly with the cytoplasm of myeloid cells. The antibody detects myeloperoxidase found in myeloid cells of acute myeloid leukemia. The myeloperoxidase gene has been localized to chromosome 17.
Order Now

SPECIFICATIONS

Antigen name Myeloperoxidase
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 59A5
Reactivity Human
Host Mouse
Isotype IgG2b kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR