Anti-E-Cadherin Mouse Monoclonal Antibody [clone: SPM471]
Supplier: Diagnostic Biosystems
MOB550-01-RUO MOB550-05-RUO MOB550-RUO
CA76634-066EA 165.22 CAD
CA76634-066 CA76634-068 CA76634-070
Anti-E-Cadherin Mouse Monoclonal Antibody [clone: SPM471]
Antibodies
E-cadherin (uvomorulin, cell-CAM120/80) is a calcium dependent cell adhesion molecule expressed predominately in epithelial tissues. It plays an important role in the growth and development of cells via the mechanisms of control of tissue architecture and the maintenance of tissue integrity. Numerous studies have demonstrated that reduction and/or loss of E-cadherin expression in carcinomas correlates positively with the potential of these tumors for invasion and metastasis.